Gene Transcripts

Gene Symbol
Grasp
Entrez Gene ID
56149
Gene Name
GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein
Gene Aliases
tamalin
Chromosome Location
Chr. 15: 101224207 - 101232756 on Build GRCm38
UniGene ID
Mm.276573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019518.3 NP_062391.3 8 1627
GenBank mRNA AB041603.1 - 8 1624
AF236099.1 - 8 1588
BC046307.1 - 8 1595 6306542

View Full Product Details