Gene Transcripts

Gene Symbol
Ick
Entrez Gene ID
56542
Gene Name
intestinal cell kinase
Gene Aliases
2210420N10Rik, AI848300, Mrk
Chromosome Location
Chr. 9: 78109192 - 78172114 on Build GRCm38
UniGene ID
Mm.288719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163780.1 NP_001157252.1 9 1449
NM_019987.2 NP_064371.2 9 1377
GenBank mRNA AF225918.1 - 9 1217
AK052178.1 - 2 346
AK087484.1 - 9 1300
AK133537.1 - 9 1316
AK154185.1 - 9 1450
AK173068.1 - 4 606
BC028863.1 - 8 1134 4224269

View Full Product Details