Gene Transcripts

Gene Symbol
Ftsj1
Entrez Gene ID
54632
Gene Name
FtsJ homolog 1 (E. coli)
Gene Aliases
AI931847, Ftsj, Ftsjl, Sfc12
Chromosome Location
Chr. X: 8238668 - 8252406 on Build GRCm38
UniGene ID
Mm.327529
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290430.1 NP_001277359.1 13 1974
NM_133991.2 NP_598752.2 13 1980
GenBank mRNA AK036313.1 - 13 1938

View Full Product Details