Gene Transcripts

Gene Symbol
Cst6
Entrez Gene ID
171096
Gene Name
cystatin E/M
Gene Aliases
Chromosome Location
Chr. 1: 220727292 - 220729000 on Build Rnor_6.0
UniGene ID
Rn.9609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133566.1 NP_598250.1 1 476
GenBank mRNA AY046415.1 - 1 476

View Full Product Details