Gene Transcripts

Gene Symbol
Trpv5
Entrez Gene ID
116469
Gene Name
transient receptor potential cation channel, subfamily V, member 5
Gene Aliases
CaT2, Ecac1
Chromosome Location
Chr. 4: 70947615 - 70974004 on Build Rnor_6.0
UniGene ID
Rn.137513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053787.2 NP_446239.2 1 248
GenBank mRNA AB032019.1 - 1 248
AF209196.1 - 199
AY762624.1 - 1 83

View Full Product Details