Gene Transcripts

Gene Symbol
ECD
Entrez Gene ID
11319
Gene Name
ecdysoneless cell cycle regulator
Gene Aliases
GCR2, HSGT1, SGT1
Chromosome Location
Chr. 10: 73134524 - 73168095 on Build GRCh38
UniGene ID
Hs.631822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135752.1 NP_001129224.1 2 328
NM_001135753.1 NP_001129225.1 2 328
NM_007265.2 NP_009196.1 2 328
GenBank mRNA AK022616.1 - 2 276
AK315711.1 - 2 301
BC000721.2 - 2 292 3504956
CR457407.1 - 1 85
D88208.1 - 2 292
DA162516.1 - 2 304

View Full Product Details