Gene Transcripts

Gene Symbol
Grasp
Entrez Gene ID
192254
Gene Name
GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein
Gene Aliases
Chromosome Location
Chr. 7: 142869785 - 142877551 on Build Rnor_6.0
UniGene ID
Rn.229849
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138894.1 NP_620249.1 1 237
GenBank mRNA AF374272.1 - 1 237

View Full Product Details