Gene Transcripts

Gene Symbol
Fosl2
Entrez Gene ID
25446
Gene Name
fos-like antigen 2
Gene Aliases
Fra2
Chromosome Location
Chr. 6: 25598936 - 25616995 on Build Rnor_6.0
UniGene ID
Rn.10962
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012954.1 NP_037086.1 1, 2 107
GenBank mRNA AY622611.1 - 1, 2 107
U18913.1 - 1, 2 107

View Full Product Details