Gene Transcripts
- Gene Symbol
- Otc
- Entrez Gene ID
- 25611
- Gene Name
- ornithine carbamoyltransferase
- Gene Aliases
- Chromosome Location
- Chr. X: 13524804 - 13601074 on Build Rnor_6.0
- UniGene ID
- Rn.2391
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013078.1 |
NP_037210.1
|
2, 3 |
319 |
|
GenBank mRNA |
BC088158.1 |
- |
2, 3 |
285 |
|
FQ211098.1 |
- |
2, 3 |
303 |
|
FQ219225.1 |
- |
2, 3 |
306 |
|
FQ219657.1 |
- |
2, 3 |
282 |
|
K00001.1 |
- |
2, 3 |
305 |
|
K03040.1 |
- |
2, 3 |
277 |
|
M11266.1 |
- |
2, 3 |
319 |
|
X01178.1 |
- |
2, 3 |
319 |
|
X01976.1 |
- |
2, 3 |
305 |
|
View Full Product Details