Gene Transcripts
- Gene Symbol
- Dio3
- Entrez Gene ID
- 29475
- Gene Name
- deiodinase, iodothyronine, type III
- Gene Aliases
- 5DIII, DIOIII
- Chromosome Location
- Chr. 6: 134627278 - 134629138 on Build Rnor_6.0
- UniGene ID
- Rn.11237
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017210.3 |
NP_058906.3
|
1 |
636 |
|
GenBank mRNA |
U24282.1 |
- |
1 |
605 |
|
View Full Product Details