Gene Transcripts
- Gene Symbol
- Ndst1
- Entrez Gene ID
- 29633
- Gene Name
- N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1
- Gene Aliases
- Hsst
- Chromosome Location
- Chr. 18: 55951497 - 55992883 on Build Rnor_6.0
- UniGene ID
- Rn.9705
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024361.1 |
NP_077337.1
|
3, 4 |
1462 |
|
GenBank mRNA |
M92042.1 |
- |
3, 4 |
1462 |
|
View Full Product Details