Gene Transcripts

Gene Symbol
Iapp
Entrez Gene ID
24476
Gene Name
islet amyloid polypeptide
Gene Aliases
Chromosome Location
Chr. 4: 176510203 - 176517903 on Build Rnor_6.0
UniGene ID
Rn.11394
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012586.2 NP_036718.1 2 178
XM_006237614.2 XP_006237676.1 3 707
GenBank mRNA J04544.1 - 3 265
M25390.1 - 3 214

View Full Product Details