Gene Transcripts

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
Chromosome Location
Chr. 5: 66596348 - 67169595 on Build GRCh38
UniGene ID
Hs.595458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164664.1 NP_001158136.1 2 720
NM_001290228.1 NP_001277157.1 2 720
NM_198828.2 NP_942123.1 2 720
XM_011543382.2 XP_011541684.1 2 701
XM_017009447.1 XP_016864936.1 2 701
XM_017009448.1 XP_016864937.1 2 701
GenBank mRNA AY439222.1 - 2 412
BC033215.1 - 2 688 5015614
CB044105.1 - 2 620

View Full Product Details