Gene Transcripts

Gene Symbol
FDX1
Entrez Gene ID
2230
Gene Name
ferredoxin 1
Gene Aliases
ADX, FDX, LOH11CR1D
Chromosome Location
Chr. 11: 110429937 - 110464884 on Build GRCh38
UniGene ID
Hs.744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004109.4 NP_004100.1 4 2010
GenBank mRNA BC010284.1 - 4 1979

View Full Product Details