Gene Transcripts

Gene Symbol
FAM35A
Entrez Gene ID
54537
Gene Name
family with sequence similarity 35 member A
Gene Aliases
FAM35A1, bA163M19.1
Chromosome Location
Chr. 10: 87094478 - 87191468 on Build GRCh38
UniGene ID
Hs.500419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019054.2 NP_061927.2 5 1896
XM_005269920.3 XP_005269977.1 5 839
XM_006717903.3 XP_006717966.1 6 2030
XM_006717905.3 XP_006717968.1 5 1953
XM_011539876.2 XP_011538178.1 4 1931
XM_011539877.2 XP_011538179.1 7 2095
XM_011539878.2 XP_011538180.1 5 1953
XM_011539879.2 XP_011538181.1 4 1939
XM_011539881.2 XP_011538183.1 4 1931
XM_011539882.2 XP_011538184.1 4 1931
XM_011539883.2 XP_011538185.1 4 884
XM_017016349.1 XP_016871838.1 9 2339
XM_017016350.1 XP_016871839.1 6 2298
XM_017016351.1 XP_016871840.1 5 1886
XM_017016352.1 XP_016871841.1 5 1886
XM_017016353.1 XP_016871842.1 5 839
GenBank mRNA AF131775.1 - 2 198
AK022978.1 - 5 1896
AK025606.1 - 3 745
BC051863.2 - 4 1842 6146593

View Full Product Details