Gene Transcripts

Gene Symbol
Esrra
Entrez Gene ID
293701
Gene Name
estrogen related receptor, alpha
Gene Aliases
ERRalpha, Errra
Chromosome Location
Chr. 1: 222168729 - 222178838 on Build Rnor_6.0
UniGene ID
Rn.130171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008511.2 NP_001008511.2 6 1095
XM_006230755.1 XP_006230817.1 6 1208
GenBank mRNA AY280663.1 - 5 989
BC104701.1 - 6 1095

View Full Product Details