Gene Transcripts

Gene Symbol
Ier2
Entrez Gene ID
494344
Gene Name
immediate early response 2
Gene Aliases
Chromosome Location
Chr. 19: 25774146 - 25775670 on Build Rnor_6.0
UniGene ID
Rn.4298
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009541.1 NP_001009541.1 1 1456
GenBank mRNA BC061717.1 - 1 1456
BC158596.1 - 2 1401

View Full Product Details