Gene Transcripts

Gene Symbol
Adamts7
Entrez Gene ID
108153
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 7
Gene Aliases
ADAM-TS7, ADAMTS7B
Chromosome Location
Chr. 9: 90162978 - 90200102 on Build GRCm38
UniGene ID
Mm.102141
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003911.2 NP_001003911.2 13, 14 2228
GenBank mRNA AY551090.1 - 13, 14 1961
BC058991.1 - 10, 11 1206
BC076575.1 - 12, 13 1936
BC141173.1 - 13, 14 2004 9055785
BC145491.1 - 13, 14 2004 9054054
CB246893.1 - 4, 5 487

View Full Product Details