Gene Transcripts

Gene Symbol
Tspan5
Entrez Gene ID
362048
Gene Name
tetraspanin 5
Gene Aliases
Tm4sf9, Tspan-5
Chromosome Location
Chr. 2: 244058186 - 244224059 on Build Rnor_6.0
UniGene ID
Rn.98240
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004090.2 NP_001004090.2 6, 7 635
GenBank mRNA AF540877.1 - 6, 7 661
BC100135.1 - 6, 7 635 7315329

View Full Product Details