Gene Transcripts

Gene Symbol
FRG2
Entrez Gene ID
448831
Gene Name
FSHD region gene 2
Gene Aliases
FRG2A
Chromosome Location
Chr. 4: 190024367 - 190027257 on Build GRCh38
UniGene ID
Hs.626907
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005217.1 NP_001005217.1 4 566
NM_001286820.1 NP_001273749.1 4 569
GenBank mRNA AY714545.1 - 4 566
BC144572.1 - 4 554 9053101

Gene Symbol
FRG2B
Entrez Gene ID
441581
Gene Name
FSHD region gene 2 family member B
Gene Aliases
Chromosome Location
Chr. 10: 133625099 - 133626795 on Build GRCh38
UniGene ID
Hs.690471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080998.1 NP_001074467.1 4 566
GenBank mRNA AY744466.1 - 4 566

View Full Product Details