Gene Transcripts

Gene Symbol
PLCXD3
Entrez Gene ID
345557
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 3
Gene Aliases
Chromosome Location
Chr. 5: 41306946 - 41510628 on Build GRCh38
UniGene ID
Hs.145404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005473.2 NP_001005473.1 3 2853
GenBank mRNA BX648329.1 - 4 2664

View Full Product Details