Gene Transcripts

Gene Symbol
OR2A5
Entrez Gene ID
393046
Gene Name
olfactory receptor family 2 subfamily A member 5
Gene Aliases
OR2A11P, OR2A26, OR2A8, OR7-138, OR7-141
Chromosome Location
Chr. 7: 144050402 - 144051337 on Build GRCh38
UniGene ID
Hs.553793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012365.1 NP_036497.1 1 257
GenBank mRNA BC136858.1 - 1 291
BC136860.1 - 1 291 9020851

View Full Product Details