Gene Transcripts

Gene Symbol
Ndufv2
Entrez Gene ID
81728
Gene Name
NADH dehydrogenase (ubiquinone) flavoprotein 2
Gene Aliases
Chromosome Location
Chr. 9: 113875718 - 113900169 on Build Rnor_6.0
UniGene ID
Rn.28882
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031064.2 NP_112326.1 6 609
GenBank mRNA BC058495.1 - 6 613
FQ215131.1 - 3 282
M22756.1 - 6 534

View Full Product Details