Gene Transcripts

Gene Symbol
Olr752
Entrez Gene ID
296004
Gene Name
olfactory receptor 752
Gene Aliases
ratchr3-96790713-96791645_ORF
Chromosome Location
Chr. 3: 102529184 - 102530116 on Build Rnor_6.0
UniGene ID
Rn.142771
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000367.1 NP_001000367.1 1 921

View Full Product Details