Gene Transcripts

Gene Symbol
Pfdn6
Entrez Gene ID
309629
Gene Name
prefoldin subunit 6
Gene Aliases
Ke2
Chromosome Location
Chr. 20: 5455974 - 5457448 on Build Rnor_6.0
UniGene ID
Rn.109
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164718.1 NP_001158190.1 5 555
NM_212506.2 NP_997671.1 4 664

View Full Product Details