Gene Transcripts
- Gene Symbol
- MYC
- Entrez Gene ID
- 4609
- Gene Name
- v-myc avian myelocytomatosis viral oncogene homolog
- Gene Aliases
- MRTL, MYCC, bHLHe39, c-Myc
- Chromosome Location
- Chr. 8: 127736069 - 127741434 on Build GRCh38
- UniGene ID
- Hs.202453
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M13929.1 |
- |
1 |
33 |
|
M13930.1 |
- |
1 |
118 |
|
View Full Product Details