Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011990.2 | NP_036120.1 | 2 | 668 | |
GenBank mRNA | AB022345.1 | - | 2 | 635 | |
AK037490.1 | - | 2 | 669 | ||
AK037742.1 | - | 2 | 643 | ||
AK089202.1 | - | 2 | 641 | ||
AK162233.1 | - | 2 | 441 | ||
AK163168.1 | - | 2 | 642 | ||
AK165599.1 | - | 2 | 644 | ||
AK171192.1 | - | 2 | 666 | ||
AK171252.1 | - | 2 | 642 | ||
AY766236.1 | - | 2 | 635 | ||
AY766237.1 | - | 635 | |||
BC141402.1 | - | 2 | 565 | 9056015 | |
BC141408.1 | - | 2 | 565 | 9056021 |