Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_028393.1 | 6 | 1361 | ||
NR_028394.1 | 7 | 1489 | |||
NR_028395.1 | 7 | 1489 | |||
NR_028396.1 | 6 | 1361 | |||
NR_028397.1 | 6 | 1226 | |||
NR_028398.1 | 4 | 980 | |||
NR_028399.1 | 8 | 1596 | |||
NR_028400.1 | 7 | 1468 |