Gene Transcripts

Gene Symbol
UBE2Q1
Entrez Gene ID
55585
Gene Name
ubiquitin conjugating enzyme E2 Q1
Gene Aliases
GTAP, NICE-5, PRO3094, UBE2Q
Chromosome Location
Chr. 1: 154548575 - 154558644 on Build GRCh38
UniGene ID
Hs.607928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017582.6 NP_060052.3 13 2896
GenBank mRNA AK056388.1 - 9 2196
AY112698.1 - 13 2870

View Full Product Details