Gene Transcripts

Gene Symbol
Raly
Entrez Gene ID
296301
Gene Name
RALY heterogeneous nuclear ribonucleoprotein
Gene Aliases
Chromosome Location
Chr. 3: 150322834 - 150387345 on Build Rnor_6.0
UniGene ID
Rn.162714
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011958.1 NP_001011958.1 6 678
XM_008762314.1 XP_008760536.1 7 1116
XM_008762315.1 XP_008760537.1 6 834
XM_008762316.1 XP_008760538.1 6 746
XM_008762317.1 XP_008760539.1 5 782
GenBank mRNA BC087074.1 - 6 678

View Full Product Details