Gene Transcripts

Gene Symbol
Txnl4b
Entrez Gene ID
292008
Gene Name
thioredoxin-like 4B
Gene Aliases
RGD1305127
Chromosome Location
Chr. 19: 42101081 - 42110222 on Build Rnor_6.0
UniGene ID
Rn.7305
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013891.1 NP_001013913.1 3 1421
XM_006255579.2 XP_006255641.1 4 1531
GenBank mRNA BC089962.1 - 3 1421

View Full Product Details