Gene Transcripts

Gene Symbol
IFITM5
Entrez Gene ID
387733
Gene Name
interferon induced transmembrane protein 5
Gene Aliases
BRIL, DSPA1, Hrmp1, OI5, fragilis4
Chromosome Location
Chr. 11: 298200 - 299526 on Build GRCh38
UniGene ID
Hs.443469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025295.2 NP_001020466.1 1 147
GenBank mRNA BC150562.1 - 1 140
BC150563.1 - 1 140 40127611
CR747200.1 - 1 149

View Full Product Details