Gene Transcripts

Gene Symbol
Tas2r115
Entrez Gene ID
353325
Gene Name
taste receptor, type 2, member 115
Gene Aliases
T2R15, Tas2r15, mGR15, mt2r49
Chromosome Location
Chr. 6: 132737054 - 132737986 on Build GRCm38
UniGene ID
Mm.377892
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207020.1 NP_996903.1 1 102
GenBank mRNA BC138393.1 - 1 151
BC138398.1 - 1 151 8861418

View Full Product Details