Gene Transcripts

Gene Symbol
OR2T8
Entrez Gene ID
343172
Gene Name
olfactory receptor family 2 subfamily T member 8
Gene Aliases
OR2T8P
Chromosome Location
Chr. 1: 247914138 - 247923152 on Build GRCh38
UniGene ID
Hs.521114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005522.1 NP_001005522.1 1 22
XM_011544178.2 XP_011542480.1 2 1365
XM_017001172.1 XP_016856661.1 2 4144

View Full Product Details