Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014061.1 | NP_001014083.1 | 971 | ||
GenBank mRNA | BC079135.1 | - | 971 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006237685.2 | XP_006237747.1 | 8 | 2306 | |
XM_006237686.2 | XP_006237748.1 | 8 | 1376 | ||
XM_006237687.2 | XP_006237749.1 | 7 | 1298 | ||
XM_008763455.1 | XP_008761677.1 | 9 | 2384 | ||
XM_008763456.1 | XP_008761678.1 | 9 | 3758 |