Gene Transcripts

Gene Symbol
Pelp1
Entrez Gene ID
360552
Gene Name
proline, glutamate and leucine rich protein 1
Gene Aliases
RGD1306320
Chromosome Location
Chr. 10: 56988076 - 57005111 on Build Rnor_6.0
UniGene ID
Rn.11628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024270.2 NP_001019441.2 12 1299
GenBank mRNA AY970831.2 - 12 1297
BC101890.1 - 12 1299 7930679

View Full Product Details