Gene Transcripts

Gene Symbol
Olfr998
Entrez Gene ID
258428
Gene Name
olfactory receptor 998
Gene Aliases
MOR175-5
Chromosome Location
Chr. 2: 85590510 - 85591521 on Build GRCm38
UniGene ID
Mm.377496
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146436.2 NP_666647.2 1 479
GenBank mRNA BC150719.1 - 1 479

View Full Product Details