Gene Transcripts

Gene Symbol
Tma7
Entrez Gene ID
66167
Gene Name
translational machinery associated 7 homolog (S. cerevisiae)
Gene Aliases
1110017O22Rik, Ccdc72
Chromosome Location
Chr. 9: 109077988 - 109082381 on Build GRCm38
UniGene ID
Mm.2395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183250.2 NP_899073.1 4 512
GenBank mRNA AK003762.1 - 4 513
AK169860.1 - 4 506
BC054363.1 - 4 484

View Full Product Details