Gene Transcripts

Gene Symbol
Olr1549
Entrez Gene ID
288196
Gene Name
olfactory receptor 1549
Gene Aliases
ratchr11-42246372-42247295_ORF
Chromosome Location
Chr. 11: 43486904 - 43487827 on Build Rnor_6.0
UniGene ID
Rn.142260
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000052.1 NP_001000052.1 1 631

View Full Product Details