Gene Transcripts

Gene Symbol
TMEM57
Entrez Gene ID
55219
Gene Name
transmembrane protein 57
Gene Aliases
MACOILIN
Chromosome Location
Chr. 1: 25430858 - 25500209 on Build GRCh38
UniGene ID
Hs.189782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018202.5 NP_060672.2 6 1344
XM_005245931.2 XP_005245988.1 6 1305
XM_011541704.2 XP_011540006.1 6 1378
GenBank mRNA AK001609.1 - 6 1255
AY845015.1 - 6 1274
BC032427.1 - 6 1212 5246967

View Full Product Details