Gene Transcripts

Gene Symbol
CASC1
Entrez Gene ID
55259
Gene Name
cancer susceptibility candidate 1
Gene Aliases
LAS1, PPP1R54
Chromosome Location
Chr. 12: 25107057 - 25195177 on Build GRCh38
UniGene ID
Hs.407771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082972.2 NP_001076441.1 8, 9 971
NM_001082973.2 NP_001076442.1 7, 8 763
NM_001204101.2 NP_001191030.1 5, 6 586
NM_001204102.2 NP_001191031.1 9, 10 819
NM_001319977.1 NP_001306906.1 6, 7 671
NM_001319978.1 NP_001306907.1 7, 8 937
NM_018272.4 NP_060742.3 8, 9 781
XM_005253406.1 XP_005253463.1 8, 9 895
XM_005253408.1 XP_005253465.1 7, 8 657
XM_005253409.1 XP_005253466.1 6, 7 671
XM_006719101.1 XP_006719164.1 8, 9 895
XM_011520723.1 XP_011519025.1 8, 9 895
XM_011520726.1 XP_011519028.1 8, 9 895
XM_011520727.1 XP_011519029.1 8, 9 895
XM_017019560.1 XP_016875049.1 7, 8 860
XM_017019561.1 XP_016875050.1 6, 7 867
XM_017019562.1 XP_016875051.1 7, 8 745
XM_017019563.1 XP_016875052.1 8, 9 777
XM_017019564.1 XP_016875053.1 7, 8 745
XM_017019565.1 XP_016875054.1 8, 9 895
XM_017019566.1 XP_016875055.1 8, 9 1106
XM_017019567.1 XP_016875056.1 5, 6 550
XM_017019568.1 XP_016875057.1 6, 7 689
GenBank mRNA AK001783.1 - 2, 3 186
AK093102.1 - 8, 9 749
AK297998.1 - 5, 6 550
AK302235.1 - 7, 8 727
AK304602.1 - 6, 7 671
AY423543.1 - 6, 7 679
BC028415.1 - 8, 9 912 4823620
BC031047.1 - 8, 9 762
BC047415.1 - 9, 10 817 5269578
BC057795.1 - 7, 8 761 5266518
BC117281.1 - 5, 6 596
BC117283.1 - 5, 6 596

View Full Product Details