Gene Transcripts

Gene Symbol
ANKRD33B
Entrez Gene ID
651746
Gene Name
ankyrin repeat domain 33B
Gene Aliases
Chromosome Location
Chr. 5: 10564323 - 10657816 on Build GRCh38
UniGene ID
Hs.26039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164440.1 NP_001157912.1 4 912
XM_005248343.4 XP_005248400.1 3 357
XM_005248344.3 XP_005248401.1 3 347

View Full Product Details