Gene Transcripts
- Gene Symbol
- CARMN
- Entrez Gene ID
- 728264
- Gene Name
- cardiac mesoderm enhancer-associated non-coding RNA
- Gene Aliases
- CARMEN, MIR143HG
- Chromosome Location
- Chr. 5: 149406845 - 149432836 on Build GRCh38
- UniGene ID
- Hs.519666
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124798.1 |
- |
1 |
740 |
|
AK126541.1 |
- |
1 |
3847 |
|
BX640700.1 |
- |
2 |
2126 |
|
View Full Product Details