Gene Transcripts
- Gene Symbol
- KIAA1257
- Entrez Gene ID
- 57501
- Gene Name
- KIAA1257
- Gene Aliases
- Chromosome Location
- Chr. 3: 128910186 - 129032431 on Build GRCh38
- UniGene ID
- Hs.518247
- Species
- Homo sapiens
- Gene Symbol
- LOC100132731
- Entrez Gene ID
- 100132731
- Gene Name
- uncharacterized LOC100132731
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.518247
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056377.1 |
- |
3 |
1422 |
|
View Full Product Details