Gene Transcripts
- Gene Symbol
- PLEKHA2
- Entrez Gene ID
- 59339
- Gene Name
- pleckstrin homology domain containing A2
- Gene Aliases
- TAPP2
- Chromosome Location
- Chr. 8: 38901235 - 38973912 on Build GRCh38
- UniGene ID
- Hs.369123
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021623.1 |
NP_067636.1
|
3, 4 |
427 |
|
XM_011544605.2 |
XP_011542907.1
|
3, 4 |
312 |
|
XM_011544606.2 |
XP_011542908.1
|
3, 4 |
363 |
|
XM_011544607.2 |
XP_011542909.1
|
3, 4 |
325 |
|
XM_017013718.1 |
XP_016869207.1
|
3, 4 |
325 |
|
XM_017013719.1 |
XP_016869208.1
|
2, 3 |
148 |
|
GenBank mRNA |
AF286164.1 |
- |
3, 4 |
352 |
|
AK098787.1 |
- |
3, 4 |
428 |
|
AK291842.1 |
- |
3, 4 |
371 |
|
AK296068.1 |
- |
2, 3 |
220 |
|
View Full Product Details