Gene Transcripts

Gene Symbol
THSD7B
Entrez Gene ID
80731
Gene Name
thrombospondin type 1 domain containing 7B
Gene Aliases
Chromosome Location
Chr. 2: 136765562 - 137677712 on Build GRCh38
UniGene ID
Hs.68533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316349.1 NP_001303278.1 7 1783
GenBank mRNA AB051466.1 - 5 1407
BC019344.1 - 5 864
BC033125.1 - 5 864

View Full Product Details