Gene Transcripts

Gene Symbol
KRT24
Entrez Gene ID
192666
Gene Name
keratin 24
Gene Aliases
K24, KA24
Chromosome Location
Chr. 17: 40694246 - 40703750 on Build GRCh38
UniGene ID
Hs.87383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019016.2 NP_061889.2 1 417
XM_017024299.1 XP_016879788.1 1 402
GenBank mRNA AK000268.1 - 1 417
BC111968.1 - 1 392 8327436

View Full Product Details