Gene Transcripts

Gene Symbol
FAM21C
Entrez Gene ID
253725
Gene Name
family with sequence similarity 21 member C
Gene Aliases
FAM21A, VPEF
Chromosome Location
Chr. 10: 45727170 - 45792964 on Build GRCh38
UniGene ID
Hs.365286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011539566.1 XP_011537868.1 27 2914
XM_011539567.1 XP_011537869.1 26 2842
XM_011539568.1 XP_011537870.1 26 2827
XM_011539569.1 XP_011537871.1 26 2761
XM_011539570.2 XP_011537872.1 26 2803
XM_011539573.2 XP_011537875.1 9 1040
XM_017016014.1 XP_016871503.1 26 2791
XM_017016017.1 XP_016871506.1 25 2755
XM_017016019.1 XP_016871508.1 25 2689
XM_017016021.1 XP_016871510.1 27 2842
XM_017016024.1 XP_016871513.1 15 1656
XM_017016025.1 XP_016871514.1 16 1727
GenBank mRNA AK001545.1 - 9 1108
AK293929.1 - 22 2398
AL050279.1 - 1 260

Gene Symbol
FAM21A
Entrez Gene ID
387680
Gene Name
family with sequence similarity 21 member A
Gene Aliases
FAM21B, bA56A21.1, bA98I6.1
Chromosome Location
Chr. 10: 50067888 - 50133509 on Build GRCh38
UniGene ID
Hs.365286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005751.2 NP_001005751.1 27 2947
XM_005269806.3 XP_005269863.2 26 2823
XM_005269807.3 XP_005269864.2 26 2808
XM_005269810.3 XP_005269867.2 9 1040
XM_006717831.3 XP_006717894.2 26 2742
XM_017016227.1 XP_016871716.1 26 2772
XM_017016230.1 XP_016871719.1 26 2760
XM_017016233.1 XP_016871722.1 24 2470
XM_017016234.1 XP_016871723.1 26 2830
XM_017016235.1 XP_016871724.1 18 1846
GenBank mRNA AK001686.1 - 24 2599
AK304116.1 - 27 2883
BC082258.1 - 27 2874 6278433
CN314884.1 - 4 272

View Full Product Details