Gene Transcripts

Gene Symbol
Pqlc3
Entrez Gene ID
298906
Gene Name
PQ loop repeat containing 3
Gene Aliases
RGD1306818
Chromosome Location
Chr. 6: 42290678 - 42302577 on Build Rnor_6.0
UniGene ID
Rn.12743
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034952.1 NP_001030124.1 7 1282
GenBank mRNA BC081879.1 - 5 2437

View Full Product Details