Gene Transcripts

Gene Symbol
Olfr328
Entrez Gene ID
258495
Gene Name
olfactory receptor 328
Gene Aliases
MOR275-2
Chromosome Location
Chr. 11: 58551305 - 58552237 on Build GRCm38
UniGene ID
Mm.339498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146502.2 NP_666713.2 1 416
GenBank mRNA BC110465.2 - 1 416
BC110466.2 - 1 416 40040839

View Full Product Details